Tisk…

3-Nitro-4-picoline, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Methyl-3-nitropyridine

M-Clarity™ quality level = MQ100

532592-1G 532592-5G
SIAL532592-1GEA 2510 CZK
SIAL532592-1G SIAL532592-5G
3-Nitro-4-picoline, Sigma-Aldrich®
3-Nitro-4-picoline
Vzorec: C₆H₆N₂O₂
MW: 138,13 g/mol
MDL Number: MFCD00051829
CAS číslo: 5832-44-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR