Tisk…

6-Amino-3-cyano-2-methylpyridine 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 6-Amino-2-methylnicotinonitrile, 6-Amino-3-cyano-2-methylpyridine, 6-Amino-2-methylpyridine-3-carbonitrile

H26000.03 H26000.06
H26000.03EA 2150 CZK
H26000.03 H26000.06
6-Amino-3-cyano-2-methylpyridine 97%
6-Amino-3-cyano-2-methylpyridine

Purity: 97% CAS No.: 183428-90-2 Molecular Formula: C7H7N3 Formula Weight: 133.15 Melting Point: 164-166° UN No.: 3439 MDL No.: MFCD05865116

Vzorec: C₇H₇N₃
MW: 133,15 g/mol
Bod tání: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD05865116
CAS číslo: 183428-90-2
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR