Tisk…

2-Acetyl-6-bromopyridine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

597872-5G 597872-1G
SIAL597872-5GEA 9280 CZK
SIAL597872-5G SIAL597872-1G
2-Acetyl-6-bromopyridine, Sigma-Aldrich®
2-Acetyl-6-bromopyridine
Vzorec: C₇H₆BrNO
MW: 200,03 g/mol
MDL Number: MFCD00272200
CAS číslo: 49669-13-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR