Tisk…
1-(6-Methoxy-2-naphthyl)ethanol ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms: DL-6-Methoxy-α-methyl-2-naphthalenemethanol, 1-(6-Methoxynaphthalen-2-yl)ethanol
B25628.06 B25628.14
B25628.06EA 1700 CZK
B25628.06 B25628.14
1-(6-Methoxy-2-naphthyl)ethanol ≥98%
1-(6-Methoxy-2-naphthyl)ethanol
Purity: 98% CAS No.: 77301-42-9 Molecular Formula: C13H14O2 Formula Weight: 202.25 Melting Point: 111-112° MDL No.: MFCD01632581
Vzorec: C₁₃H₁₄O₂
MW: 202.25 g/mol
Bod tání: 111…112 °C
Storage Temperature: Ambient
MDL Number: MFCD01632581
CAS číslo: 77301-42-9