1-Acetyl-4-fluoronaphthalene ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4'-Fluoro-1'-acetonaphthone, 1-(4-Fluoronaphthalen-1-yl)ethanone, 1-(4-Fluoro-1-naphthyl)ethanone
B25004.03
B25004.14
B25004.03EA
1460
CZK
B25004.03
B25004.14
1-Acetyl-4-fluoronaphthalene ≥97%
1-Acetyl-4-fluoronaphthalene
Purity: 97%
CAS No.: 316-68-7
Molecular Formula: C12H9FO
Formula Weight: 188.20
Flash Point:
Vzorec:
C₁₂H₉FO MW: 188,20 g/mol Bod varu: 164…166 °C (14 mmHg) Hustota: 1,21 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00134475 CAS číslo: 316-68-7 EINECS: 206-261-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...