Tisk…
1-Nitroso-2-naphthol-3,6-disulphonic acid disodium salt ≥90%
Dodavatel: Thermo Scientific
Varování
Synonyms: 3-Hydroxy-4-nitroso-2,7-naphthalenedisulphonic acid disodium salt, Nitroso-R salt
B23375.14 B23375.22 B23375.36
B23375.14EA 1650 CZK
B23375.14 B23375.22 B23375.36
1-Nitroso-2-naphthol-3,6-disulphonic acid disodium salt ≥90%
1-Nitroso-2-naphthol-3,6-disulphonic acid disodium salt
Purity: 90+% CAS No.: 525-05-3 Molecular Formula: C10H5NNa2O8S2 Formula Weight: 377.26 Melting Point: 300° Beilstein Reference: 3643930 Merck Reference: 14,6644 MDL No.: MFCD00003885
Vzorec: C₁₀H₅NNa₂O₈S₂
MW: 377,26 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003885
CAS číslo: 525-05-3
EINECS: 208-369-1
Merck Index: 14,6644