Manganese(II) wolframate ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Manganese(II) tungsten oxide
39374.18
39374.30
39374.18EA
4600
CZK
39374.18
39374.30
Manganese(II) wolframate ≥99.9% (metals basis)
Manganese(II) wolframate
Purity: 99.9%
CAS No.: 14177-46-9
Molecular Formula: MnWO4
Formula Weight: 302.79
Physical Form: -200 Mesh Powder
Odour: Odourless
MDL No.: MFCD00014212
Vzorec:
MnO₄W MW: 302.79 g/mol Hustota: 7.23 Storage Temperature: Ambient |
MDL Number:
MFCD00014212 CAS číslo: 14177-46-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...