Tisk…

Tungsten(VI) oxychloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

265012-25G 265012-5G
SIAL265012-25GEA 18410 CZK
SIAL265012-25G SIAL265012-5G
Tungsten(VI) oxychloride, Sigma-Aldrich®
Tungsten(VI) oxychloride
Vzorec: Cl₄OW
MDL Number: MFCD00011465
CAS číslo: 13520-78-0
EINECS: 236-863-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR