Sodium phenoxide ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Sodium phenylate
B25002.18
B25002.30
B25002.18EA
2700
CZK
B25002.18
B25002.30
Sodium phenoxide ≥98%
Sodium phenoxide
Purity: 98%
CAS No.: 139-02-6
Molecular Formula: C6H5NaO
Formula Weight: 116.09
Odour: Phenol-like
Melting Point:
Vzorec:
C₆H₅NaO MW: 116,09 g/mol Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013134 CAS číslo: 139-02-6 EINECS: 205-347-3 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...