Tisk…

4-Amino-7-(trifluoromethyl)quinoline 98%

Dodavatel: Thermo Fisher Scientific
Varování

H50355.03 H50355.MD
H50355.03EA 4740 CZK
H50355.03 H50355.MD
4-Amino-7-(trifluoromethyl)quinoline 98%
4-Amino-7-(trifluoromethyl)quinoline

Purity: 98% CAS No.: 243666-11-7 Molecular Formula: C10H7F3N2 Formula Weight: 212.18 MDL No.: MFCD00269411

Vzorec: C₁₀H₇F₃N₂
Storage Temperature: Ambient
MDL Number: MFCD00269411
CAS číslo: 243666-11-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR