Tisk…

6-Fluoroquinoline ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H35900.MD H35900.03
H35900.MDEA 1610 CZK
H35900.MD H35900.03
6-Fluoroquinoline ≥97%
6-Fluoroquinoline
Vzorec: C₉H₆FN
MW: 147.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01685512
CAS číslo: 396-30-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR