2,4-Dimethyl-6-iodoaniline ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Iodo-4,6-dimethylaniline
H51875.03
H51875.06
H51875.03EA
2720
CZK
H51875.03
H51875.06
2,4-Dimethyl-6-iodoaniline ≥98%
2,4-Dimethyl-6-iodoaniline
Purity: 98%
CAS No.: 4102-54-9
Molecular Formula: C8H10IN
Formula Weight: 247.08
Melting Point: 64-65°
UN No.: 2811
MDL No.: MFCD07779005
Vzorec:
C₈H₁₀IN MW: 247,08 g/mol Bod tání: 64…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD07779005 CAS číslo: 4102-54-9 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...