Tisk…

N-Cbz-D-phenylalaninol ≥98%

Dodavatel: Thermo Fisher Scientific

H27819.06 H27819.03
H27819.06EA 3400 CZK
H27819.06 H27819.03
N-Cbz-D-phenylalaninol ≥98%
N-Cbz-D-phenylalaninol

Purity: 98% CAS No.: 58917-85-4 Molecular Formula: C17H19NO3 Formula Weight: 285.35 Melting Point: 92-95° Optical Rotation: 42° (c=1 in methanol)

Vzorec: C₁₇H₁₉NO₃
Bod tání: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00191193
CAS číslo: 58917-85-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR