Tisk…

Methyl-4-(hydroxymethyl)benzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 4-(hydroxymethyl)benzoate

M-Clarity™ quality level = MQ100

266477-10G 266477-1G
SIAL266477-10GEA 3470 CZK
SIAL266477-10G SIAL266477-1G
Methyl-4-(hydroxymethyl)benzoate, Sigma-Aldrich®
Methyl-4-(hydroxymethyl)benzoate
Vzorec: C₉H₁₀O₃
MW: 166,18 g/mol
MDL Number: MFCD00009984
CAS číslo: 6908-41-4
EINECS: 230-017-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR