Tisk…

Methyl-3-methylsalicylate ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

B21341.04 B21341.09
B21341.04EA 1310 CZK
B21341.04 B21341.09
Methyl-3-methylsalicylate ≥97%
Methyl-3-methylsalicylate

Purity: 97% CAS No.: 23287-26-5 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 28-30° Boiling Point: 234-236° MDL No.: MFCD00020051

Vzorec: C₉H₁₀O₃
Bod varu: 234…236 °C
Bod tání: 28…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00020051
CAS číslo: 23287-26-5
EINECS: 245-557-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR