4-Methyl-3-nitrobenzamide ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L18270.06
L18270.14
L18270.06EA
560
CZK
L18270.06
L18270.14
4-Methyl-3-nitrobenzamide ≥98%
4-Methyl-3-nitrobenzamide
Purity: 98%
CAS No.: 19013-11-7
Molecular Formula: C8H8N2O3
Formula Weight: 180.16
Melting Point: 164-167°
Beilstein Reference: 2102644
Vzorec:
C₈H₈N₂O₃ Bod tání: 164…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00027392 CAS číslo: 19013-11-7 EINECS: 242-752-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...