Tisk…

N-Boc-D-phenylalaninol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Boc-D-Phenylalaninol

M-Clarity™ quality level = MQ100

479594-1G 479594-5G
SIAL479594-1GEA 2080 CZK
SIAL479594-1G SIAL479594-5G
N-Boc-D-phenylalaninol, Sigma-Aldrich®
N-Boc-D-phenylalaninol
Vzorec: C₁₄H₂₁NO₃
MW: 251,3 g/mol
MDL Number: MFCD00216472
CAS číslo: 106454-69-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR