Tisk…

N-Boc-D-phenylalaninol ≥98%

Dodavatel: Thermo Scientific

Synonyms: Boc-D-Phenylalaninol

H27713.03 H27713.06
H27713.03EA 1880 CZK
H27713.03 H27713.06
N-Boc-D-phenylalaninol ≥98%
N-Boc-D-phenylalaninol

Purity: 98% CAS No.: 106454-69-7 Molecular Formula: C14H21NO3 Formula Weight: 251.33 Melting Point: 94-96° MDL No.: MFCD00216472

Vzorec: C₁₄H₂₁NO₃
MW: 251,3 g/mol
Bod tání: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00216472
CAS číslo: 106454-69-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR