Tisk…

(1H-Imidazol-4-yl)methanol hydrochloride ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-Imidazolemethanol hydrochloride, 4(5)-Imidazolemethanol hydrochloride, 4-(Hydroxymethyl)imidazole hydrochloride

L20182.06 L20182.14
L20182.06EA 2530 CZK
L20182.06 L20182.14
(1H-Imidazol-4-yl)methanol hydrochloride ≥98%
(1H-Imidazol-4-yl)methanol hydrochloride

Purity: 98+% CAS No.: 32673-41-9 Molecular Formula: C4H6N2O.HCl Formula Weight: 134.57 Storage Temperature: 2°C to 8°C Melting Point: 104-108° Beilstein Reference: 3562041 MDL No.: MFCD00012697

Vzorec: C₄H₇ClN₂O
MW: 134,57 g/mol
Bod tání: 104…108 °C
MDL Number: MFCD00012697
CAS číslo: 32673-41-9
EINECS: 251-150-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR