Tisk…

4-(Chlorosulphonyl)benzoic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

252700-1G 252700-25G 252700-5G
SIAL252700-1GEA 882 CZK
SIAL252700-1G SIAL252700-25G SIAL252700-5G
4-(Chlorosulphonyl)benzoic acid, Sigma-Aldrich®
4-(Chlorosulphonyl)benzoic acid
Vzorec: C₇H₅ClO₄S
MW: 220,63 g/mol
MDL Number: MFCD00007448
CAS číslo: 10130-89-9
EINECS: 233-367-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR