N-Boc-aniline 97%
Dodavatel: Thermo Fisher Scientific
H55232.06
H55232.14
H55232.06EA
1600
CZK
H55232.06
H55232.14
N-Boc-aniline 97%
N-Boc-aniline
Vzorec:
C₁₁H₁₅NO₂ |
MDL Number:
MFCD00043723 CAS číslo: 3422-01-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...