2-Amino-5-chlorobenzenesulphonic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B25738.06
B25738.14
B25738.06EA
2220
CZK
B25738.06
B25738.14
2-Amino-5-chlorobenzenesulphonic acid ≥98%
2-Amino-5-chlorobenzenesulphonic acid
Purity: 98%
CAS No.: 133-74-4
Molecular Formula: C6H6ClNO3S
Formula Weight: 207.64
Melting Point:
Vzorec:
C₆H₆ClNO₃S MW: 207.64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00035776 CAS číslo: 133-74-4 EINECS: 205-119-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...