3-(Boc-amino)phenylboronic acid pinacol ester 97%
Dodavatel: Thermo Scientific
Synonyms:
tert-Butyl (3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate, 3-(Boc-amino)benzeneboronic acid pinacol ester
H27026.06
H27026.03
H27026.06EA
8170
CZK
H27026.06
H27026.03
3-(Boc-amino)phenylboronic acid pinacol ester 97%
3-(Boc-amino)phenylboronic acid pinacol ester
Purity: 97%
CAS No.: 330793-09-4
Molecular Formula: C17H26BNO4
Formula Weight: 319.21
Melting Point: 118-120°
MDL No.: MFCD03789256
Vzorec:
C₁₇H₂₆BNO₄ MW: 319,21 g/mol Bod tání: 118…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD03789256 CAS číslo: 330793-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...