Diallyl maleate 95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 95%
CAS No.: 999-21-3
Molecular Formula: C10H12O4
Formula Weight: 196.21
Flash Point: 130°(266°F)
Refractive Index: 1.4670
Melting Point: -47°
Boiling Point: 109-110°/3mm
Beilstein Reference: 1725954
UN No.: 2810
MDL No.: MFCD00010763
Vzorec:
C₁₀H₁₂O₄ Bod varu: 109…110 °C (3 mmHg) Bod tání: –47 °C Hustota: 1,072 Bod vzplanutí: 130 °C (266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010763 CAS číslo: 999-21-3 EINECS: 213-658-0 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...