Tisk…

(R)-(-)-2-Fluoro-α-methyl-4-biphenylacetic acid

Dodavatel: Cayman Chemical

70255-100 70255-500
CAYM70255-100EA 1320 CZK
CAYM70255-100 CAYM70255-500
(R)-(-)-2-Fluoro-α-methyl-4-biphenylacetic acid
(R)-(-)-2-Fluoro-α-methyl-4-biphenylacetic acid
Vzorec: C₁₅H₁₃FO₂
MDL Number: MFCD00869714
CAS číslo: 51543-40-9
EINECS: 257-264-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR