N-Boc-L-tyrosine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
BOC-L-Tyrosine, Boc-Tyr-OH
A10810.03
A10810.06
A10810.14
A10810.03EA
618
CZK
A10810.03
A10810.06
A10810.14
N-Boc-L-tyrosine ≥98%
N-Boc-L-tyrosine
Purity: 98+%
CAS No.: 3978-80-1
Molecular Formula: C14H19NO5
Formula Weight: 281.31
Storage Temperature: 2°C to 8°C
Melting Point: 141° dec.
Beilstein Reference: 2816406
Optical Rotation: +37° (c=1 in dioxan)
MDL No.: MFCD00037179
Vzorec:
C₁₄H₁₉NO₅ |
MDL Number:
MFCD00037179 CAS číslo: 3978-80-1 EINECS: 223-613-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...