Tisk…

Sodium 4-aminohippurate hydrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

A3759-100G A3759-25G
SIALA3759-100GEA 32260 CZK
SIALA3759-100G SIALA3759-25G
Sodium 4-aminohippurate hydrate, Sigma-Aldrich®
Sodium 4-aminohippurate hydrate
Vzorec: C₉H₉N₂NaO₃·H₂O
MW: 216,17 g/mol (anhy)
MDL Number: MFCD00044689
CAS číslo: 94-16-6
EINECS: 202-309-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR