Tisk…

2-(Methylthio)ethylamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(Methylthio)ethanamine

M-Clarity™ quality level = MQ100

632929-1G 632929-5G
SIAL632929-1GEA 2400 CZK
SIAL632929-1G SIAL632929-5G
2-(Methylthio)ethylamine, Sigma-Aldrich®
2-(Methylthio)ethylamine
Vzorec: C₃H₉NS
MW: 91,18 g/mol
MDL Number: MFCD00014825
CAS číslo: 18542-42-2
EINECS: 242-412-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR