N,N'-Diacetyl-1,6-diaminohexane, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
224235-10G
224235-50G
SIAL224235-10GEA
0
CZK
SIAL224235-10G
SIAL224235-50G
N,N'-Diacetyl-1,6-diaminohexane, Sigma-Aldrich®
N,N'-Diacetyl-1,6-diaminohexane
Vzorec:
C₁₀H₂₀N₂O₂ |
MDL Number:
MFCD00008684 CAS číslo: 3073-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...