N-Boc-L-alaninol ≥99%
Dodavatel: Thermo Fisher Scientific
B22399.06
B22399.14
B22399.06EA
3750
CZK
B22399.06
B22399.14
N-Boc-L-alaninol ≥99%
N-Boc-L-alaninol
Purity: 99%
CAS No.: 79069-13-9
Molecular Formula: C8H17NO3
Formula Weight: 175.23
Storage Temperature: 2°C to 8°C
Melting Point: 60-62°
Beilstein Reference: 3648840
Optical Rotation: -8.6° (c=1.2 in methanol)
MDL No.: MFCD00043121
Vzorec:
C₈H₁₇NO₃ Bod tání: 60…62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00043121 CAS číslo: 79069-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...