Tisk…

1-Ethoxy-1-(tributylstannyl)ethene, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

275123-100G 275123-1G 275123-25G 275123-5G
SIAL275123-100GEA 101450 CZK
SIAL275123-100G SIAL275123-1G SIAL275123-25G SIAL275123-5G
1-Ethoxy-1-(tributylstannyl)ethene, Sigma-Aldrich®
1-Ethoxy-1-(tributylstannyl)ethene
Vzorec: C₁₆H₃₄OSn
MDL Number: MFCD00010240
CAS číslo: 97674-02-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR