Tributyltin hydride 1 mol/l in cyclohexane, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
704091-10ML
704091-50ML
SIAL704091-10MLEA
0
CZK
SIAL704091-10ML
SIAL704091-50ML
Tributyltin hydride 1 mol/l in cyclohexane, Sigma-Aldrich®
Tributyltin hydride
Vzorec:
C₁₂H₂₈Sn MW: 291,06 g/mol |
MDL Number:
MFCD00009416 CAS číslo: 688-73-3 EINECS: 211-704-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...