4,6-Dichloro-2-(methylthio)pyrimidine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
144533-25G
144533-5G
SIAL144533-25GEA
9190
CZK
SIAL144533-25G
SIAL144533-5G
4,6-Dichloro-2-(methylthio)pyrimidine, Sigma-Aldrich®
4,6-Dichloro-2-(methylthio)pyrimidine
Vzorec:
C₅H₄Cl₂N₂S MW: 195,07 g/mol |
MDL Number:
MFCD00006086 CAS číslo: 6299-25-8 EINECS: 228-577-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...