3-Amino-5-(methylthio)-1H-pyrazole-4-carbonitrile 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-Amino-5-(methylthio)pyrazole-4-carbonitrile
B20991.03
B20991.06
B20991.03EA
2410
CZK
B20991.03
B20991.06
3-Amino-5-(methylthio)-1H-pyrazole-4-carbonitrile 97%
3-Amino-5-(methylthio)-1H-pyrazole-4-carbonitrile
Purity: 97%
CAS No.: 72760-85-1
Molecular Formula: C5H6N4S
Formula Weight: 154.19
Melting Point: 148-150°
UN No.: 3439
MDL No.: MFCD00052745
Vzorec:
C₅H₆N₄S MW: 154,19 g/mol Bod tání: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052745 CAS číslo: 72760-85-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...