Tisk…

(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-Boc-1H-pyrazole-4-boronic acid, 1-tert-Butoxycarbonyl-1H-pyrazole-4-boronic acid, 1H-Pyrazole-4-boronic acid, N1-BOC protected

715263-500MG 715263-1G
SIAL715263-500MGEA 4540 CZK
SIAL715263-500MG SIAL715263-1G
(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid, Sigma-Aldrich®
(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid
Vzorec: C₈H₁₃BN₂O₄
MW: 212,01 g/mol
MDL Number: MFCD09951915
CAS číslo: 947533-31-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR