(S)-3-(Aminomethyl)-1-boc-piperidine 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
tert-Butyl (S)-3-(aminomethyl)piperidine-1-carboxylate, (S)-1-BOC-3-(Aminomethyl)piperidine
H52822.03
H52822.06
H52822.03EA
3850
CZK
H52822.03
H52822.06
(S)-3-(Aminomethyl)-1-boc-piperidine 97%
(S)-3-(Aminomethyl)-1-boc-piperidine
Purity: 97%
CAS No.: 140645-24-5
Molecular Formula: C11H22N2O2
Formula Weight: 214.31
MDL No.: MFCD03839877
Vzorec:
C₁₁H₂₂N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03839877 CAS číslo: 140645-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...