Tisk…

α-Methylcinnamic acid ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: 2-Methyl-3-phenyl-2-propenoic acid

L00806.06 L00806.14 L00806.22
L00806.06EA 848 CZK
L00806.06 L00806.14 L00806.22
α-Methylcinnamic acid ≥99%
α-Methylcinnamic acid

Purity: 99% CAS No.: 1199-77-5 Molecular Formula: C10H10O2 Formula Weight: 162.19 Melting Point: 78-82° Boiling Point: 288° MDL No.: MFCD00002652

Vzorec: C₁₀H₁₀O₂
MW: 162.19 g/mol
Bod varu: 288 °C
Bod tání: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00002652
CAS číslo: 1199-77-5
EINECS: 214-847-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR