3-(2-Aminoethylamino)propylmethyldimethoxysilane 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-(2-Aminoethylamino)propyldimethoxymethylsilane
B23159.14
B23159.22
B23159.36
B23159.14EA
814
CZK
B23159.14
B23159.22
B23159.36
3-(2-Aminoethylamino)propylmethyldimethoxysilane 97%
3-(2-Aminoethylamino)propylmethyldimethoxysilane
Purity: 97%
CAS No.: 3069-29-2
Molecular Formula: (CH3O)2(CH3)Si(CH2)3NHCH2CH2NH2
Formula Weight: 206.36
Flash Point: 90°(194°F)
Refractive Index: 1.450
Odour: Amine-like
Boiling Point: 265°
Beilstein Reference: 2074259
UN No.: 2735
MDL No.: MFCD00054776
Vzorec:
C₈H₂₂N₂O₂Si MW: 206,36 g/mol Bod varu: 265 °C Hustota: 0,968 Bod vzplanutí: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00054776 CAS číslo: 3069-29-2 EINECS: 221-336-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...