Tisk…

(3-Aminopropyl)diethoxymethylsilane 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 3-(Diethoxy(methyl)silyl)propan-1-amine

L16401.14 L16401.22
L16401.14EA 1040 CZK
L16401.14 L16401.22
(3-Aminopropyl)diethoxymethylsilane 97%
(3-Aminopropyl)diethoxymethylsilane

Purity: 97% CAS No.: 3179-76-8 Molecular Formula: C8H21NO2Si Formula Weight: 191.35 Flash Point: 85°(185°F) Refractive Index: 1.4270 Melting Point: -20° Boiling Point: 197-199° Beilstein Reference: 1744264 UN No.: 2735 MDL No.: MFCD00039785

Vzorec: C₈H₂₁NO₂Si
MW: 191,35 g/mol
Bod varu: 197…199 °C
Bod tání: < –20 °C
Hustota: 0,920
Bod vzplanutí: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039785
CAS číslo: 3179-76-8
EINECS: 221-660-8
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR