Tisk…

1-Boc-3-methylpiperazine ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H52809.06 H52809.14
H52809.06EA 2800 CZK
H52809.06 H52809.14
1-Boc-3-methylpiperazine ≥97%
1-Boc-3-methylpiperazine

Purity: 97% CAS No.: 120737-59-9 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 MDL No.: MFCD03001706

Vzorec: C₁₀H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03001706
CAS číslo: 120737-59-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR