Tisk…

(R)-(+)-1-Boc-3-methylpiperazine ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (3R)-3-Methylpiperazine, N1-BOC protected, (R)-1-Boc-3-Methylpiperazine, (R)-1-Boc-3-methyl-piperazine

H27350.06 H27350.03
H27350.06EA 3780 CZK
H27350.06 H27350.03
(R)-(+)-1-Boc-3-methylpiperazine ≥97%
(R)-(+)-1-Boc-3-methylpiperazine

Purity: 97% CAS No.: 163765-44-4 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 Melting Point: 40-45° Optical Rotation: +16° (c=1 in dioxane)

Vzorec: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Bod tání: 40…45 °C
Storage Temperature: Ambient
MDL Number: MFCD02683205
CAS číslo: 163765-44-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR