Tisk…

4,4',4''-Trimethyltrityl alcohol ≥98%

Dodavatel: Thermo Fisher Scientific

L02392.06 L02392.14
L02392.06EA 3250 CZK
L02392.06 L02392.14
4,4',4''-Trimethyltrityl alcohol ≥98%
4,4',4''-Trimethyltrityl alcohol

Purity: 98+% CAS No.: 3247-00-5 Molecular Formula: C22H22O Formula Weight: 302.42 Melting Point: 92-96° Beilstein Reference: 2055803

Vzorec: C₂₂H₂₂O
Bod tání: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00014919
CAS číslo: 3247-00-5
EINECS: 221-822-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR