Tisk…

(S)-(+)-2-(Methoxymethyl)pyrrolidine ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (S)-2-(Methoxymethyl)pyrrolidine

L09608.03 L09608.06
L09608.03EA 2490 CZK
L09608.03 L09608.06
(S)-(+)-2-(Methoxymethyl)pyrrolidine ≥98%
(S)-(+)-2-(Methoxymethyl)pyrrolidine

Purity: 98% CAS No.: 63126-47-6 Molecular Formula: C6H13NO Formula Weight: 115.18 Flash Point: 45°(113°F) Refractive Index: 1.4457 Boiling Point: 61-62°/40mm Beilstein Reference: 3587324 Optical Rotation: +2.2° (c=2 in benzene) UN No.: 1993 MDL No.: MFCD00010408

Vzorec: C₆H₁₃NO
MW: 115.18 g/mol
Bod varu: 61…62 °C (40 mmHg)
Hustota: 0.933
Bod vzplanutí: 45 °C (113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010408
CAS číslo: 63126-47-6
EINECS: 626-212-9
UN: 1993
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR