(S)-1-Boc-piperidine-3-carboxylic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(S)-Boc-nipecotic acid, 1-Boc-L-nipecotic acid, (S)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid
H52575.06
H52575.14
H52575.06EA
3250
CZK
H52575.06
H52575.14
(S)-1-Boc-piperidine-3-carboxylic acid ≥97%
(S)-1-Boc-piperidine-3-carboxylic acid
Purity: 97%
CAS No.: 88495-54-9
Molecular Formula: C11H19NO4
Formula Weight: 229.28
Melting Point: 165-169°
Optical Rotation: +57° (c=1 in chloroform)
Vzorec:
C₁₁H₁₉NO₄ MW: 229,28 g/mol Bod tání: 165…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179172 CAS číslo: 88495-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...