4,4-Difluoropiperidine hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4,4-Difluoropiperidine HCl
M-Clarity™ quality level = MQ100
665525-1G
SIAL665525-1GEA
3020
CZK
SIAL665525-1G
4,4-Difluoropiperidine hydrochloride, Sigma-Aldrich®
4,4-Difluoropiperidine hydrochloride
Vzorec:
C₅H₁₀ClF₂N MW: 157.59 g/mol |
MDL Number:
MFCD03095381 CAS číslo: 144230-52-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...