Tisk…

(R)-(-)-3-Fluoropyrrolidine hydrochloride ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (R)-(-)-3-Fluoropyrrolidine Hydrochloride

H26947.06 H26947.03
H26947.06EA 20280 CZK
H26947.06 H26947.03
(R)-(-)-3-Fluoropyrrolidine hydrochloride ≥97%
(R)-(-)-3-Fluoropyrrolidine hydrochloride

Purity: 97% CAS No.: 136725-55-8 Molecular Formula: C4H8FN.HCl Formula Weight: 125.57 Melting Point: 181-184° MDL No.: MFCD04038717

Vzorec: C₄H₉ClFN
MW: 125.57 g/mol
Bod tání: 181…184 °C
Storage Temperature: Ambient
MDL Number: MFCD04038717
CAS číslo: 136725-55-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR