Tisk…

4-(4-Piperidinyloxy)benzonitrile ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

H50958.03 H50958.MD
H50958.03EA 8410 CZK
H50958.03 H50958.MD
4-(4-Piperidinyloxy)benzonitrile ≥98%
4-(4-Piperidinyloxy)benzonitrile

Purity: 98% CAS No.: 224178-67-0 Molecular Formula: C12H14N2O Formula Weight: 202.26 UN No.: 3439 MDL No.: MFCD07368391

Vzorec: C₁₂H₁₄N₂O
MW: 202,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368391
CAS číslo: 224178-67-0
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR