Tisk…

Tetrazolium violet ≥98%

Dodavatel: Thermo Scientific

A16008.03 A16008.06 A16008.14
A16008.03EA 1700 CZK
A16008.03 A16008.06 A16008.14
Tetrazolium violet ≥98%
Tetrazolium violet

Purity: 98+% CAS No.: 1719-71-7 Molecular Formula: C23H17ClN4 Formula Weight: 384.87 Melting Point: 247-249° MDL No.: MFCD00011875

Vzorec: C₂₃H₁₇ClN₄
MW: 384,87 g/mol
Bod tání: 247…249 °C
Storage Temperature: Ambient
MDL Number: MFCD00011875
CAS číslo: 1719-71-7
EINECS: 217-008-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR