Tisk…

Bathocuproin ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline

B22841.03 B22841.06
B22841.03EA 3040 CZK
B22841.03 B22841.06
Bathocuproin ≥98%
Bathocuproin

Purity: 98% CAS No.: 4733-39-5 Molecular Formula: C26H20N2 Formula Weight: 360.46 Melting Point: 279-285° Beilstein Reference: 306714

Vzorec: C₂₆H₂₀N₂
MW: 360,46 g/mol
Bod tání: 279…285 °C
Storage Temperature: Ambient
MDL Number: MFCD00004972
CAS číslo: 4733-39-5
EINECS: 225-240-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR