Pilocarpine nitrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
P0597-25G
P0597-5G
SIALP0597-25GEA
0
CZK
SIALP0597-25G
SIALP0597-5G
Pilocarpine nitrate, Sigma-Aldrich®
Pilocarpine nitrate
Vzorec:
C₁₁H₁₇N₃O₅ |
MDL Number:
MFCD00078497 CAS číslo: 148-72-1 EINECS: 205-723-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...